Assay Details
| Gene Symbol: SEC16A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011519251.1 | XP_011517553.1 | 28 - 29 | 7106 | 127 | |
| XM_011519253.1 | XP_011517555.1 | 28 - 29 | 7097 | 127 | ||
| XM_011519255.2 | XP_011517557.1 | 28 - 29 | 7040 | 127 | ||
| XM_011519263.2 | XP_011517565.1 | 29 - 30 | 6329 | 127 | ||
| GenBank mRNA | AK074565.1 | - | 4 - 5 | 468 | 127 | |
| AK301371.1 | - | 6 - 7 | 507 | 127 | ||
| AK302210.1 | - | 5 - 6 | 500 | 127 | ||
| AK316194.1 | - | 4 - 5 | 402 | 127 | ||
| BC028183.1 | - | 24 - 25 | 3044 | 127 | ||
| BC125019.1 | - | 28 - 29 | 5038 | 127 | ||
More Information
Set Membership: |
|

SEC16A
Genomic Map