Assay Details
Gene Symbol: ABCA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080284.2 | NP_525023.2 | 18 - 19 | 2557 | 137 | |
XM_011524574.2 | XP_011522876.1 | 12 - 13 | 1885 | 137 | ||
XM_017024404.1 | XP_016879893.1 | 18 - 19 | 2869 | 137 | ||
XM_017024405.1 | XP_016879894.1 | 18 - 19 | 2869 | 137 | ||
XM_017024406.1 | XP_016879895.1 | 14 - 15 | 1837 | 137 | ||
XM_017024407.1 | XP_016879896.1 | 15 - 16 | 1941 | 137 | ||
XM_017024408.1 | XP_016879897.1 | 4 - 5 | 430 | 137 | ||
GenBank mRNA | AF373290.1 | - | 17 - 18 | 2385 | 137 | |
AY028898.1 | - | 18 - 19 | 2557 | 137 | ||
BC125231.1 | - | 18 - 19 | 2556 | 137 | ||
BC125232.1 | - | 18 - 19 | 2556 | 137 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17001827.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
