Assay Details
Gene Symbol: NR3C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204265.1 | NP_001191194.1 | 7 | 3962 | 72 | |
GenBank mRNA | AA737284.1 | - | 1 | 72 | ||
U25029.1 | - | 1 | 1448 | 72 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Rho GTPase activating protein 26 | ARHGAP26 |
GRAF, GRAF1, OPHN1L, OPHN1L1 |
-64.9kb | Anti-sense | Chr. 5 - 142770370 - 143229007 on NCBI Build GRCh38 |
Downstream | microRNA 5197 | MIR5197 | 384.4kb | Anti-sense | Chr. 5 - 143679860 - 143679971 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
