Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000431144.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000431144.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000431144.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000431144.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000440578.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000440578.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000440578.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000448942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000448942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000448942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000448942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000448942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000448942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000448942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000448942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000448942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000448942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000448942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000448942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000448942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000448942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000448942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000448942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000606327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000606327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000606327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000606998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000606998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000606998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000606998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000606998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000606998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000607671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000607671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000607671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000607671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000607671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000607671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
ENST00000607671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
NR_049793.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
NR_049793.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
NR_049793.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
NR_049793.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
NR_049793.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013316.hg.1 |
NR_049793.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000431144.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000431144.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000431144.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000431144.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000440578.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000440578.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000440578.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000448942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000448942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000448942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000448942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000448942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000448942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000448942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000448942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000448942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000448942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000448942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000448942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000448942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000448942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000448942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000448942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000606327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000606327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000606327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000606998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000606998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000606998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000606998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000606998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000606998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000607671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000607671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000607671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000607671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000607671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000607671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
ENST00000607671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
NR_049793.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
NR_049793.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
NR_049793.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
NR_049793.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
NR_049793.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013316.hg.1 |
NR_049793.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600192159.hg.1 |
ENST00000448942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600192159.hg.1 |
ENST00000448942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600192159.hg.1 |
ENST00000606998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600192159.hg.1 |
ENST00000607671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600192159.hg.1 |
NR_049793.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600192159.hg.1 |
ENST00000448942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600192159.hg.1 |
ENST00000448942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600192159.hg.1 |
ENST00000606998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600192159.hg.1 |
ENST00000607671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600192159.hg.1 |
NR_049793.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600192159.hg.1 |
TC0600013316.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600192158.hg.1 |
ENST00000606998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600192158.hg.1 |
NR_049793.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600192158.hg.1 |
ENST00000606998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600192158.hg.1 |
NR_049793.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600192158.hg.1 |
TC0600013316.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600192160.hg.1 |
ENST00000606998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600192160.hg.1 |
NR_049793.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600192160.hg.1 |
ENST00000606998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600192160.hg.1 |
NR_049793.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600192160.hg.1 |
TC0600013316.hg.1 |
Clariom D TC |