Assay Details
Gene Symbol: SFN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005544307.2 | XP_005544364.1 | 1 | 2533 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
