Assay Details
| Gene Symbol: Acvr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110204.1 | NP_001103674.1 | 7 - 8 | 1210 | 71 | |
| NM_001110205.1 | NP_001103675.1 | 5 - 6 | 688 | 71 | ||
| NM_007394.3 | NP_031420.2 | 6 - 7 | 965 | 71 | ||
| GenBank mRNA | AF332087.1 | - | 4 - 5 | 598 | 71 | |
| AF332088.1 | - | 4 - 5 | 598 | 71 | ||
| AK052246.1 | - | 800 | 71 | |||
| AK053428.1 | - | 7 - 8 | 1209 | 71 | ||
| AK150014.1 | - | 6 - 7 | 965 | 71 | ||
| AK150075.1 | - | 6 - 7 | 965 | 71 | ||
| BC058718.1 | - | 4 - 5 | 686 | 6418954 | 71 | |
| L15436.1 | - | 5 - 6 | 1038 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Acvr1
Genomic Map