Assay Details
Gene Symbol: Dll3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007866.2 | NP_031892.2 | 2 - 3 | 386 | 80 | |
GenBank mRNA | AB013440.1 | - | 2 - 3 | 373 | 80 | |
AK144282.1 | - | 2 - 3 | 387 | 80 | ||
BC052002.1 | - | 2 - 3 | 363 | 6404029 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
