Assay Details
Gene Symbol: Prkci | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008857.3 | NP_032883.2 | 15 - 16 | 1711 | 89 | |
GenBank mRNA | AK144979.1 | - | 15 - 16 | 1711 | 89 | |
AK147710.1 | - | 15 - 16 | 1683 | 89 | ||
AK167411.1 | - | 15 - 16 | 1680 | 89 | ||
AK220517.1 | - | 15 - 16 | 1735 | 89 | ||
BC021630.1 | - | 15 - 16 | 1582 | 3980628 | 89 | |
D28577.1 | - | 15 - 16 | 1506 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
