Assay Details
Gene Symbol: Tpd52l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009413.1 | NP_033439.1 | 1 - 2 | 208 | 55 | |
GenBank mRNA | AF004428.1 | - | 1 - 2 | 208 | 55 | |
AK131855.1 | - | 1 - 2 | 105 | 55 | ||
BC141068.1 | - | 1 - 2 | 95 | 9055680 | 55 | |
BC145321.1 | - | 1 - 2 | 95 | 9053876 | 55 | |
BC145322.1 | - | 1 - 2 | 95 | 55 | ||
BC171924.1 | - | 1 - 2 | 95 | 55 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AF004428.1,AK131855.1,BC141068.1,BC145321.1,NM_009413.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
