Assay Details
| Gene Symbol: Mocs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013826.3 | NP_038854.2 | 1 - 2 | 86 | 75 | |
| GenBank mRNA | AF091872.1 | - | 1 - 2 | 37 | 75 | |
| AK010112.1 | - | 1 - 2 | 66 | 75 | ||
| BC024371.1 | - | 1 - 2 | 46 | 4983231 | 75 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF091872.1,AK010112.1,BC024371.1,NM_013826.3 transcripts.
Set Membership: |
|

Mocs2
Genomic Map