Assay Details
Gene Symbol: Rgs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015811.2 | NP_056626.2 | 3 - 4 | 350 | 117 | |
GenBank mRNA | AF215667.1 | - | 3 - 4 | 281 | 117 | |
AK089293.1 | - | 2 - 3 | 817 | 117 | ||
AK138688.1 | - | 3 - 4 | 306 | 117 | ||
AK154394.1 | - | 306 | 117 | |||
AK170366.1 | - | 3 - 4 | 302 | 117 | ||
AK170957.1 | - | 3 - 4 | 306 | 117 | ||
AK171306.1 | - | 647 | 117 | |||
BC028634.1 | - | 3 - 4 | 281 | 1328269 | 117 | |
BC052684.1 | - | 1 - 2 | 1136 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
G-protein coupled receptor signaling pathway
regulation of G-protein coupled receptor protein signaling pathway
negative regulation of signal transduction
termination of G-protein coupled receptor signaling pathway
positive regulation of GTPase activity
GTPase activator activity
G-protein coupled receptor signaling pathway
regulation of G-protein coupled receptor protein signaling pathway
negative regulation of signal transduction
termination of G-protein coupled receptor signaling pathway
positive regulation of GTPase activity
GTPase activator activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P260683 | BC052684.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P260683 | BC028634.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P260683 | AK138688.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P260683 | AK154841.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P260683 | AK170366.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P260683 | BC052684.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P260683 | BC028634.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P260683 | AK138688.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P260683 | AK154841.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P260683 | AK170366.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 262380 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 603205 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 723785 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 10980 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 819520 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 49158 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P144994 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P260683 | BC052684.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P260683 | BC028634.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P260683 | AK138688.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P260683 | AK154841.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P260683 | AK170366.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P260683 | BC052684.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P260683 | BC028634.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P260683 | AK138688.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P260683 | AK154841.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P260683 | AK170366.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P260683 | BC052684.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P260683 | BC028634.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P260683 | AK138688.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P260683 | AK154841.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P260683 | AK170366.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P260683 | BC052684.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P260683 | BC028634.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P260683 | AK138688.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P260683 | AK154841.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P260683 | AK170366.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20682517 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20682528 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20682516 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20682520 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20682529 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20682507 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20682538 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20682531 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20682542 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20682543 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20682541 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20682540 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20682536 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20682525 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20682522 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20682549 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20682535 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20682523 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20682509 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20682521 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20682513 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20682547 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20682545 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20682532 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20682533 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20682519 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20682527 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20682511 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P144994 | - | - |
Whole Mouse Genome G4122A | A_51_P260683 | BC052684.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P260683 | BC028634.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P260683 | AK138688.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P260683 | AK154841.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P260683 | AK170366.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P260683 | BC052684.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P260683 | BC028634.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P260683 | AK138688.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P260683 | AK154841.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P260683 | AK170366.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110580 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P144994 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01109361 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00581997 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00582002 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01109358 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01109365 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00582007 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682513 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682564 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682478 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682488 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682566 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682455 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682529 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682560 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682464 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682511 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682494 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682543 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682542 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682480 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682525 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682538 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682459 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682448 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682471 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682528 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682496 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682470 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682556 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682516 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682522 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682554 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682474 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682492 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682483 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682517 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682452 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682527 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682497 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682485 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682533 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682473 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682519 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682481 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682503 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682536 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682520 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682499 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682466 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682507 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682509 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682547 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682532 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682558 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682472 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682505 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682541 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682453 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682531 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682451 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682450 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682523 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682445 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682456 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682521 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682501 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20682562 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682499 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682511 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682513 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682523 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682522 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682558 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682532 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682490 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682562 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682533 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682555 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682483 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682541 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00582004 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682504 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682556 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00945462 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682560 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00418582 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682478 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682519 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682516 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682509 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682529 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682494 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682485 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682492 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682472 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682481 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682484 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682497 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682528 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682520 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682471 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00582005 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682503 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01028632 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682553 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682547 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00582012 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682554 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682531 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682501 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682564 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01028631 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682542 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00945477 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682566 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682507 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01109359 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682488 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682517 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682538 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682515 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682474 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00653661 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20682552 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00418584 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10358413 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10358411 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10358409 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10358412 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10358414 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10358410 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1417601_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10358408 | - | - |
Mouse Genome 430 2.0 Array | 1417601_at | - | - |
Mouse Genome 430 2.0 Array | 1417601_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 50778_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 50778_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0100026053.mm.1 | NM_015811.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100026053.mm.1 | NM_015811.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100026053.mm.1 | TC0100003207.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100026055.mm.1 | NM_015811.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100026055.mm.1 | NM_015811.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100026055.mm.1 | TC0100003207.mm.1 | MTA TC |
Mouse Expression Set 430 | 1417601_PM_at | - | - |
Mouse Expression Set 430 | 1417601_at | - | - |
Mouse Genome 430A 2.0 Array | 1417601_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1417601_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417601_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417601_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 923224 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 4540181 | - | - |
MouseWG-6 v2 Expression BeadChip | 4060347 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 4060347 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 106040215 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 4540181 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004540181 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004060347 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2596596 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2625377 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2482308 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2596596 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2625377 | - | - |
mouse-6 v1.1 (Illumina) | scl16187.2_8-S | - | - |
mouse-6 v1.1 (Illumina) | scl000798.1_9-S | - | - |
mouse-6 v1.1 (Illumina) | scl0050778.2_103-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4060347 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 106040215 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4540181 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl000798.1_9-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0050778.2_103-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4060347 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106040215 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4540181 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2596596 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2625377 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2897891 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2596596 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2625377 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2897891 | - | - |
