Assay Details
| Gene Symbol: Rexo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024233.3 | NP_077195.2 | 3 - 4 | 364 | 69 | |
| GenBank mRNA | AK007731.1 | - | 3 - 4 | 366 | 69 | |
| AK150287.1 | - | 3 - 4 | 280 | 69 | ||
| AK171619.1 | - | 3 - 4 | 267 | 69 | ||
| AK172641.1 | - | 3 - 4 | 342 | 69 | ||
| AK206506.1 | - | 1 - 2 | 50 | 69 | ||
| AK207467.1 | - | 1 - 2 | 50 | 69 | ||
| AK208282.1 | - | 1 - 2 | 50 | 69 | ||
| AK210585.1 | - | 1 - 2 | 50 | 69 | ||
| AK214375.1 | - | 1 - 2 | 50 | 70 | ||
| BC003445.1 | - | 3 - 4 | 300 | 2651110 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rexo2
Genomic Map