Assay Details
Gene Symbol: Raf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029780.3 | NP_084056.1 | 10 - 11 | 1532 | 73 | |
GenBank mRNA | AB057655.1 | - | 9 - 10 | 1077 | 73 | |
AB057663.2 | - | 10 - 11 | 1302 | 73 | ||
AK018272.1 | - | 1 - 2 | 88 | 73 | ||
AK141745.1 | - | 10 - 11 | 1533 | 73 | ||
BC015273.1 | - | 10 - 11 | 1411 | 4222659 | 73 | |
BC056439.1 | - | 10 - 11 | 1449 | 73 | ||
BC092040.1 | - | 10 - 11 | 1397 | 2645313 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
