Assay Details
Gene Symbol: Ebpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026598.3 | NP_080874.2 | 2 - 3 | 287 | 95 | |
GenBank mRNA | AF243434.2 | - | 2 - 3 | 243 | 95 | |
AK011237.1 | - | 2 - 3 | 246 | 95 | ||
AK019140.1 | - | 2 - 3 | 139 | 95 | ||
AK019963.1 | - | 2 - 3 | 282 | 95 | ||
AK163371.1 | - | 2 - 3 | 246 | 95 | ||
BC027422.1 | - | 2 - 3 | 265 | 4035224 | 95 | |
BY349880.1 | - | 2 - 3 | 290 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
