Assay Details
Gene Symbol: Endou | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168693.1 | NP_001162164.1 | 7 - 8 | 1002 | 61 | |
NM_008902.3 | NP_032928.1 | 6 - 7 | 1001 | 61 | ||
GenBank mRNA | AK169457.1 | - | 5 - 6 | 1092 | 61 | |
M95545.1 | - | 6 - 7 | 1001 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
