Assay Details
| Gene Symbol: Gkn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025466.1 | NP_079742.1 | 4 - 5 | 362 | 93 | |
| GenBank mRNA | AK008622.1 | - | 4 - 5 | 362 | 93 | |
| AK008640.1 | - | 4 - 5 | 360 | 93 | ||
| AK008641.1 | - | 4 - 5 | 362 | 93 | ||
| AK008647.1 | - | 4 - 5 | 359 | 93 | ||
| AK008722.1 | - | 4 - 5 | 360 | 93 | ||
| AK008745.1 | - | 4 - 5 | 362 | 93 | ||
| AK008933.1 | - | 4 - 5 | 364 | 93 | ||
| AK008956.1 | - | 4 - 5 | 359 | 93 | ||
| AK008990.1 | - | 4 - 5 | 364 | 93 | ||
| AK009145.1 | - | 4 - 5 | 360 | 93 | ||
| AK019050.1 | - | 4 - 5 | 363 | 93 | ||
| AK028136.1 | - | 4 - 5 | 358 | 93 | ||
| AY456960.1 | - | 4 - 5 | 362 | 93 | ||
| BC099446.1 | - | 4 - 5 | 345 | 30521253 | 93 | |
| BC132540.1 | - | 4 - 5 | 352 | 40130817 | 93 | |
| BC132542.1 | - | 4 - 5 | 352 | 40130819 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gkn1
Genomic Map