Assay Details
Gene Symbol: Fiz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011813.3 | NP_035943.3 | 1 - 2 | 148 | 134 | |
GenBank mRNA | AF126747.1 | - | 1 - 2 | 106 | 134 | |
BC006633.1 | - | 1 - 2 | 148 | 3482191 | 134 | |
BC138231.1 | - | 1 - 2 | 98 | 8861250 | 134 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AF126747.1,BC006633.1,BC138231.1,NM_011813.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
