Assay Details
Gene Symbol: Surf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013677.2 | NP_038705.2 | 5 - 6 | 731 | 74 | |
NR_073420.1 | 5 - 6 | 771 | 74 | |||
GenBank mRNA | AK002469.1 | - | 5 - 6 | 545 | 74 | |
AK154726.1 | - | 5 - 6 | 551 | 74 | ||
AK208539.1 | - | 1 - 2 | 74 | 74 | ||
BC004755.1 | - | 5 - 6 | 731 | 3584373 | 74 | |
BC052500.1 | - | 5 - 6 | 718 | 5255619 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21081131 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21081132 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P332602 | AK208539.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P332602 | BC004755.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P332602 | BC052500.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P332602 | CB524655.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P332602 | NM_013677.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P332602 | AK208539.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P332602 | BC004755.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P332602 | BC052500.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P332602 | CB524655.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P332602 | NM_013677.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 284271 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 145591 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178683 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P332602 | AK208539.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P332602 | BC004755.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P332602 | BC052500.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P332602 | CB524655.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P332602 | NM_013677.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P332602 | AK208539.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P332602 | BC004755.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P332602 | BC052500.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P332602 | CB524655.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P332602 | NM_013677.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P332602 | AK208539.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P332602 | BC004755.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P332602 | BC052500.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P332602 | CB524655.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P332602 | NM_013677.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P332602 | AK208539.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P332602 | BC004755.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P332602 | BC052500.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P332602 | CB524655.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P332602 | NM_013677.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081108 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081121 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081128 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081124 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081112 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081125 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081130 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081119 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081117 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081131 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081106 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081110 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081114 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P178683 | - | - |
Whole Mouse Genome G4122A | A_51_P332602 | AK208539.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P332602 | BC004755.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P332602 | BC052500.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P332602 | CB524655.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P332602 | NM_013677.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P332602 | AK208539.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P332602 | BC004755.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P332602 | BC052500.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P332602 | CB524655.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P332602 | NM_013677.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116405 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P178683 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01852385 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01852382 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01852378 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01852373 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21081132 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21081131 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081047 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081148 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081158 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081161 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081020 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081139 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081145 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081046 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081164 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081040 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081153 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081029 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081142 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081027 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081162 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081141 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081144 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081023 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081045 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081042 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081156 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081213 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081030 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081137 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081150 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081160 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081212 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21081145 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21081141 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01582397 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21081148 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21081137 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21081160 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21081150 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21081139 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21081164 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01582440 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01582441 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21081144 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21081153 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10481139 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10481142 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10481143 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10481144 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10481138 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10481145 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10481137 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10481146 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10481141 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10470323 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10481140 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10481136 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1450561_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10481135 | - | - |
Mouse Genome 430 2.0 Array | 1450561_a_at | - | - |
Mouse Genome 430 2.0 Array | 1450561_PM_a_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20930_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20930_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0200026924.mm.1 | NM_013677.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200026924.mm.1 | NM_013677.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200026924.mm.1 | NR_073420.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200026924.mm.1 | TC0200003204.mm.1 | MTA TC |
Mouse Expression Set 430 | 1450561_PM_a_at | - | - |
Mouse Expression Set 430 | 1450561_a_at | - | - |
Murine Genome U74v2 Set | 101031_at | - | - |
Murine Genome U74v2 Set | 101031_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1450561_PM_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1450561_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450561_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450561_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 755727 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 5130048 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 5130048 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005130048 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2657141 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2657141 | - | - |
mouse-6 v1.1 (Illumina) | scl19516.4.19_7-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5130048 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl19516.4.19_7-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5130048 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2883561 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2657141 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2657141 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2883561 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl19516.4.19_7-S | - | - |
