Assay Details
| Gene Symbol: Tex261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009357.2 | NP_033383.1 | 3 - 4 | 395 | 111 | |
| GenBank mRNA | AK149789.1 | - | 3 - 4 | 360 | 111 | |
| BC003802.1 | - | 3 - 4 | 339 | 3486968 | 111 | |
| X81058.1 | - | 3 - 4 | 341 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24595446 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24595445 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24595443 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24595442 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24595444 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P242733 | BC003802.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P242733 | X80425.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P242733 | X81058.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P242733 | AK157834.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P242733 | AK170049.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 630456 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 293923 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 657829 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 240624 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608253 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 14510 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P121320 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P242733 | BC003802.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P242733 | X80425.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P242733 | X81058.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P242733 | AK157834.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P242733 | AK170049.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24595465 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24595473 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24595458 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24595426 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24595442 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24595462 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24595450 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24595449 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24595439 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24595443 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24595429 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24595470 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24595467 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24595433 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24595460 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24595436 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24595438 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24595423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24595444 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24595440 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24595446 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24595468 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24595427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24595454 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24595472 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24595431 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24595435 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24595456 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05962527 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P121320 | - | - |
| Whole Mouse Genome G4122A | A_52_P378452 | AK013969.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P378452 | AK013971.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P654021 | BC003802.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P654021 | X81058.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P654021 | AK149789.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P654021 | NM_009357.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P147654 | AK170049.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P147654 | AK013969.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P147654 | AK013971.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129436 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P121320 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05298582 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05256487 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05292001 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05298584 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24595445 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24595446 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24595443 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24595444 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24595442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24595383 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24595460 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05298602 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24595412 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24595404 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24595435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24595476 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24595477 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24595456 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05256486 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24595433 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05730233 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24595407 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05879308 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05298600 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24595458 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24595429 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24595454 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24595398 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24595472 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05298601 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24595459 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24595421 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24595445 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05879303 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24595413 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05298568 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24595401 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24595450 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05032321 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24595449 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24595415 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24595396 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24595452 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05818596 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24595436 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05879311 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05298572 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24595431 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24595463 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24595409 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05730234 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24595470 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24595473 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05425888 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24595465 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24595479 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24595402 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24595406 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24595443 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05108999 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24595442 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24595418 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10545754 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10545757 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10545756 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10545752 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10545753 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10545755 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452768_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450965_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423384_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10545751 | - | - |
| Mouse Genome 430 2.0 Array | 1452768_at | - | - |
| Mouse Genome 430 2.0 Array | 1450965_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1423384_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1452768_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1423384_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1450965_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21766_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21766_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600020090.mm.1 | TC0600002582.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600020090.mm.1 | NM_009357.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600020090.mm.1 | NM_009357.2 | Refseq |
| Mouse Expression Set 430 | 1423384_PM_s_at | - | - |
| Mouse Expression Set 430 | 1450965_at | - | - |
| Mouse Expression Set 430 | 1450965_PM_at | - | - |
| Mouse Expression Set 430 | 1423384_s_at | - | - |
| Murine Genome U74v2 Set | 96166_PM_at | - | - |
| Murine Genome U74v2 Set | 101517_at | - | - |
| Murine Genome U74v2 Set | 96166_at | - | - |
| Murine Genome U74v2 Set | 101517_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450965_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423384_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450965_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423384_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423384_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450965_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423384_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452768_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452768_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450965_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 793512 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2340037 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2630672 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002630672 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002340037 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2605494 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1253322 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1253322 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2605494 | - | - |
| mouse-6 v1.1 (Illumina) | scl28784.6_160-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0021766.1_289-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2340037 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2630672 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0021766.1_289-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl28784.6_160-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2340037 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2630672 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1253322 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2605494 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2605494 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1253322 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0021766.1_289-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl28784.6_160-S | - | - |

Tex261
Genomic Map