Assay Details
Gene Symbol: Psen1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008943.2 | NP_032969.1 | 2 - 3 | 441 | 78 | |
GenBank mRNA | AK134232.1 | - | 2 - 3 | 124 | 78 | |
AK134615.1 | - | 2 - 3 | 163 | 78 | ||
AK148101.1 | - | 2 - 3 | 162 | 78 | ||
AK165984.1 | - | 2 - 3 | 162 | 78 | ||
BC014744.1 | - | 2 - 3 | 192 | 4511140 | 78 | |
BC030409.1 | - | 2 - 3 | 116 | 5133302 | 78 | |
BC071233.1 | - | 2 - 3 | 180 | 3673609 | 78 | |
L42177.1 | - | 2 - 3 | 137 | 78 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK134232.1,AK134615.1,AK148101.1,AK165984.1,BC014744.1,BC030409.1,BC071233.1,L42177.1,NM_008943.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
