Assay Details
Gene Symbol: Psen1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008943.2 | NP_032969.1 | 6 - 7 | 1034 | 94 | |
GenBank mRNA | AK134232.1 | - | 6 - 7 | 717 | 94 | |
AK134615.1 | - | 6 - 7 | 756 | 94 | ||
AK148101.1 | - | 6 - 7 | 755 | 94 | ||
AK165984.1 | - | 6 - 7 | 755 | 94 | ||
AK169963.1 | - | 2 - 3 | 161 | 94 | ||
BC014744.1 | - | 6 - 7 | 785 | 4511140 | 94 | |
BC030409.1 | - | 6 - 7 | 709 | 5133302 | 94 | |
BC071233.1 | - | 6 - 7 | 773 | 3673609 | 94 | |
L42177.1 | - | 6 - 7 | 730 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
