Assay Details
Gene Symbol: Nt5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015807.1 | NP_056622.1 | 3 - 4 | 375 | 64 | |
GenBank mRNA | AF078840.1 | - | 3 - 4 | 375 | 64 | |
AK007418.1 | - | 3 - 4 | 414 | 64 | ||
BC024368.1 | - | 3 - 4 | 378 | 5065485 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
nucleotide metabolic process
pyrimidine deoxyribonucleotide catabolic process
deoxyribonucleotide catabolic process
dephosphorylation
nucleotide binding
protein tyrosine/serine/threonine phosphatase activity
nucleotidase activity
5'-nucleotidase activity
protein tyrosine/threonine phosphatase activity
JUN kinase phosphatase activity
phosphohistidine phosphatase activity
inositol bisphosphate phosphatase activity
hydrolase activity
phosphatase activity
MAP kinase tyrosine/serine/threonine phosphatase activity
inositol-1,3,4-trisphosphate 4-phosphatase activity
pyrimidine nucleotide binding
NADP phosphatase activity
transmembrane receptor protein phosphatase activity
inositol-1,3,4,5,6-pentakisphosphate 3-phosphatase activity
inositol-1,4,5,6-tetrakisphosphate 6-phosphatase activity
inositol-4,5-bisphosphate 5-phosphatase activity
5-amino-6-(5-phosphoribitylamino)uracil phosphatase activity
phosphatidylinositol-4-phosphate phosphatase activity
phosphatidylinositol-3,5-bisphosphate 5-phosphatase activity
metal ion binding
phosphatidylinositol-3,5-bisphosphate 3-phosphatase activity
inositol phosphate phosphatase activity
inositol-1,3,4,5,6-pentakisphosphate 1-phosphatase activity
inositol-3,4-bisphosphate 4-phosphatase activity
inositol-1,3,4-trisphosphate 1-phosphatase activity
inositol-1,3,4,6-tetrakisphosphate 6-phosphatase activity
inositol-1,3,4,6-tetrakisphosphate 1-phosphatase activity
phosphatidylinositol-1,4,5-trisphosphate 5-phosphatase activity
IDP phosphatase activity
nucleotide metabolic process
pyrimidine deoxyribonucleotide catabolic process
deoxyribonucleotide catabolic process
dephosphorylation
nucleotide binding
protein tyrosine/serine/threonine phosphatase activity
nucleotidase activity
5'-nucleotidase activity
protein tyrosine/threonine phosphatase activity
JUN kinase phosphatase activity
phosphohistidine phosphatase activity
inositol bisphosphate phosphatase activity
hydrolase activity
phosphatase activity
MAP kinase tyrosine/serine/threonine phosphatase activity
inositol-1,3,4-trisphosphate 4-phosphatase activity
pyrimidine nucleotide binding
NADP phosphatase activity
transmembrane receptor protein phosphatase activity
inositol-1,3,4,5,6-pentakisphosphate 3-phosphatase activity
inositol-1,4,5,6-tetrakisphosphate 6-phosphatase activity
inositol-4,5-bisphosphate 5-phosphatase activity
5-amino-6-(5-phosphoribitylamino)uracil phosphatase activity
phosphatidylinositol-4-phosphate phosphatase activity
phosphatidylinositol-3,5-bisphosphate 5-phosphatase activity
metal ion binding
phosphatidylinositol-3,5-bisphosphate 3-phosphatase activity
inositol phosphate phosphatase activity
inositol-1,3,4,5,6-pentakisphosphate 1-phosphatase activity
inositol-3,4-bisphosphate 4-phosphatase activity
inositol-1,3,4-trisphosphate 1-phosphatase activity
inositol-1,3,4,6-tetrakisphosphate 6-phosphatase activity
inositol-1,3,4,6-tetrakisphosphate 1-phosphatase activity
phosphatidylinositol-1,4,5-trisphosphate 5-phosphatase activity
IDP phosphatase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28153834 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28153835 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28153833 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28153836 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2038162 | AK007418.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2038162 | NM_015807.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2038162 | AF078840.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 90726 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P111031 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2038162 | AK007418.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2038162 | NM_015807.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2038162 | AF078840.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P284976 | AK007418.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P284976 | NM_015807.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P284976 | AF078840.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P423849 | AK007418.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P423849 | BC024368.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P423849 | NM_015807.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P423849 | AF078840.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28153838 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28153852 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28153840 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28153833 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28153856 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28153841 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28153837 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28153849 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28153867 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28153854 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28153827 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28153847 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28153826 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28153829 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28153824 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28153857 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28153835 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28153839 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28153869 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28153834 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28153832 | - | - |
Whole Mouse Genome G4122A | A_51_P284976 | AK007418.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P284976 | NM_015807.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P284976 | AF078840.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P423849 | AK007418.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P423849 | BC024368.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P423849 | NM_015807.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P423849 | AF078840.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100953 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09397075 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09471736 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09471735 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09471740 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09471737 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09708094 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28153836 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28153833 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28153835 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28153834 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153837 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153826 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153857 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153841 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153852 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153832 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153834 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153869 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153782 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153840 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153779 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153839 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153856 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153833 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153829 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153854 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153824 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153777 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153772 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153827 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153835 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28153849 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28153835 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28153838 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28153824 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28153844 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28153866 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28153826 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28153869 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28153840 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09314851 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28153827 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09199491 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28153837 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28153850 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09397075 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28153832 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28153854 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09015010 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28153868 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28153857 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28153829 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28153833 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09199492 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28153846 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10392941 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10392938 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10392939 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10392937 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10392940 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1417252_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10392936 | - | - |
Mouse Genome 430 2.0 Array | 1417252_at | - | - |
Mouse Genome 430 2.0 Array | 1417252_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 50773_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 50773_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100038273.mm.1 | NM_015807.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100038273.mm.1 | NM_015807.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100038273.mm.1 | TC1100004075.mm.1 | MTA TC |
Mouse Expression Set 430 | 1417252_PM_at | - | - |
Mouse Expression Set 430 | 1417252_at | - | - |
Mouse Genome 430A 2.0 Array | 1417252_at | - | - |
Mouse Genome 430A 2.0 Array | 1417252_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417252_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417252_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 681490 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 70064 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 70064 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000070064 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1223097 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1223097 | - | - |
mouse-6 v1.1 (Illumina) | scl39338.3_4-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 70064 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl39338.3_4-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 70064 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1223097 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1223097 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl39338.3_4-S | - | - |
