Assay Details
Gene Symbol: Cfap57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026789.4 | NP_081065.2 | 20 - 21 | 3406 | 73 | |
GenBank mRNA | AK003848.1 | - | 1 - 2 | 104 | 73 | |
BC062805.1 | - | 1 - 2 | 111 | 1478343 | 73 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK003848.1,BC062805.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1977787 | AK003848.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 609913 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 287165 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 813317 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 925772 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 427910 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 436339 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 43058 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 242613 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 762417 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 682692 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 42004 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P122958 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P128640 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241111 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241117 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241158 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241131 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241148 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241160 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241155 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241151 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241139 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241167 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241166 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241110 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241137 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241124 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241163 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241146 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241145 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241129 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241162 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241169 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241121 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241149 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241134 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241115 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241130 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241133 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241122 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241153 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241138 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241109 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241141 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241143 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241120 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241126 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241147 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P122958 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05374396 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P128640 | - | - |
Whole Mouse Genome G4122A | A_51_P112203 | AK003848.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139288 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P122958 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P128640 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03494313 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03494306 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03494319 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03494298 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03494324 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241177 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241126 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241180 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241059 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241038 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241142 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241170 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241004 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241005 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241096 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241063 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23240999 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241021 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241024 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241031 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241080 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241122 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241064 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241120 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241107 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241172 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241083 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241092 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241111 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23240982 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23240989 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241020 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241104 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241091 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241052 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241167 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241115 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241162 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241134 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241188 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241166 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241027 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241010 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241019 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241034 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241182 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241016 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241110 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23240983 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241046 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241100 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241077 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241029 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241139 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241192 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241145 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241041 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241069 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241084 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241105 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241194 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241009 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241179 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241065 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241116 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241007 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241175 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241138 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241089 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241067 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241141 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241033 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241153 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241131 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241098 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241044 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241158 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241101 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241147 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241048 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241124 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241056 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241160 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23240993 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241109 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241151 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241039 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241148 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23240984 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241129 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241195 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241191 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241025 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241006 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241054 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241014 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241017 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23240986 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241042 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241075 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241133 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241062 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241037 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241012 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241066 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241143 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241022 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241114 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241066 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03408725 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03767666 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03447542 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241075 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241138 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03237802 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241129 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03494324 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241143 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03322930 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241147 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241184 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03237792 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241158 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241069 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241182 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241141 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241164 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03604057 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03494306 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03237796 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03408719 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241084 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241098 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241166 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241160 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241175 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241080 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241110 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241186 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241142 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241191 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03494308 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241194 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241122 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241134 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241172 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04092132 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241120 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241089 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241101 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241097 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03767670 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241091 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241092 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04092134 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241167 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241146 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241077 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241117 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241104 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241126 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03447541 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241118 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03540576 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03832225 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241105 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241139 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03853881 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241153 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241081 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241148 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03998665 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241155 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03540563 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03494311 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241177 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241188 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241180 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241109 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241111 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241192 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241145 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241151 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241073 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23241162 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10515804 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10515807 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10515805 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10515806 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1429808_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1429869_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10515808 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10515803 | - | - |
Mouse Genome 430 2.0 Array | 1429869_at | - | - |
Mouse Genome 430 2.0 Array | 1429808_at | - | - |
Mouse Genome 430 2.0 Array | 1429869_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1429808_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68625_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68625_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0400027552.mm.1 | TC0400003396.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400027552.mm.1 | NM_026789.4 | Refseq |
Mouse Expression Set 430 | 1429808_at | - | - |
Mouse Expression Set 430 | 1429808_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1429869_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1429869_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 599563 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 100430088 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 100430088 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1770463 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001770463 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2614983 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2437650 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2614983 | - | - |
mouse-6 v1.1 (Illumina) | scl3024.1.1_249-S | - | - |
mouse-6 v1.1 (Illumina) | scl23911.2.1_11-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1770463 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 100430088 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl23911.2.1_11-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1770463 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100430088 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2614983 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2614983 | - | - |
