Assay Details
| Gene Symbol: Drap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291080.1 | NP_001278009.1 | 2 - 3 | 386 | 82 | |
| NM_024176.2 | NP_077138.1 | 2 - 3 | 386 | 82 | ||
| GenBank mRNA | AK005445.1 | - | 1 - 2 | 121 | 82 | |
| AK010152.1 | - | 2 - 3 | 340 | 82 | ||
| AK087456.1 | - | 2 - 3 | 212 | 82 | ||
| AK154251.1 | - | 1 - 2 | 166 | 82 | ||
| BC002090.1 | - | 2 - 3 | 323 | 3488139 | 82 | |
| CT010290.1 | - | 2 - 3 | 121 | 82 | ||
 Back To Top
 Back To TopMore Information
Updated Mapping Information
 This assay targets the same location as MTA transcript cluster(s) TC1900000964.mm.1
					            	This assay targets the same location as MTA transcript cluster(s) TC1900000964.mm.1| Set Membership: |  Ambion siRNA available  Amplicon length between 71 and 85  Mammalian Gene Collection  Detect Genomic DNA  Mus musculus  Inventoried  Probe spans exons | 
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
 Back To Top
 Back To Top 
                                                     
                                                     
                                                    
 Drap1
Drap1 Genomic Map
Genomic Map 
                		 Back To Top
 Back To Top