Assay Details
Gene Symbol: Riiad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025506.2 | NP_079782.1 | 5 - 6 | 392 | 75 | |
GenBank mRNA | AK009192.1 | - | 5 - 6 | 394 | 75 | |
AK019569.1 | - | 3 - 4 | 202 | 75 | ||
BC107021.1 | - | 4 - 5 | 779 | 40049310 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P183213 | NM_025506.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P183213 | AK009192.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P183213 | AK019569.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P183213 | BC107021.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P183213 | NM_025506.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P183213 | AK009192.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P183213 | AK019569.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P183213 | BC107021.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 512737 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 357974 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 634161 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 238936 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 508032 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 726445 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 61950 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P124126 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P183213 | NM_025506.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P183213 | AK009192.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P183213 | AK019569.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P183213 | BC107021.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P183213 | NM_025506.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P183213 | AK009192.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P183213 | AK019569.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P183213 | BC107021.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22339219 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22339234 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22339212 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22339229 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22339211 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22339245 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22339236 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22339231 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22339241 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22339221 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22339205 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22339204 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22339223 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22339243 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22339209 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22339249 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22339224 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22339207 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22339246 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22339227 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22339214 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22339248 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22339238 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22339228 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22339217 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P124126 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04992642 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04992661 | - | - |
Whole Mouse Genome G4122A | A_51_P183213 | NM_025506.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P183213 | AK009192.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P183213 | AK019569.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P183213 | BC107021.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P183213 | NM_025506.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P183213 | AK009192.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P183213 | AK019569.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P183213 | BC107021.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125534 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P124126 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02870035 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02298737 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02591767 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02858121 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02870026 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02858125 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02298735 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02591766 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339279 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339236 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339131 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339278 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339288 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339162 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339268 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339277 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339203 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339191 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339206 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339210 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339290 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339271 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339161 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339248 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339192 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339228 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339149 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339174 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339250 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339116 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339179 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339190 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339184 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339249 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339223 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339269 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339264 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339287 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339121 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339266 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339125 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339132 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339256 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339243 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339189 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339246 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339273 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339254 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339275 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339180 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339214 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339217 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339284 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339212 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339143 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339196 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339259 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339265 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339234 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339238 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339211 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339261 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339152 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339140 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339219 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339160 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339185 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339280 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339291 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339239 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339257 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339197 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339252 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339193 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339187 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339200 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339204 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339170 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339241 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339117 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339177 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339122 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339167 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339172 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339141 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339163 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339229 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339144 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339221 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339245 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339158 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339286 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339136 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02695518 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22339162 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22339170 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22339203 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22339223 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22339191 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22339202 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02863903 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22339167 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22339205 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22339206 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22339158 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22339190 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02963214 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22339217 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22339161 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22339224 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22339195 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22339209 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22339189 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02407331 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22339204 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02468322 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22339200 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22339128 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22339186 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22339210 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22339181 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03169424 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03085032 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22339219 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02963209 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22339221 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22339225 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22339163 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22339180 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22339174 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02858130 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22339193 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22339129 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22339130 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02402612 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22339179 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02695517 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22339183 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22339192 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02462716 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02863905 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22339185 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22339211 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10499989 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10499991 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10499994 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10499995 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10499992 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10499990 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10499993 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1429419_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10499988 | - | - |
Mouse Genome 430 2.0 Array | 1429419_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1429419_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66353_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66353_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0300018750.mm.1 | NM_025506.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300018750.mm.1 | NM_025506.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300018750.mm.1 | TC0300002444.mm.1 | MTA TC |
Mouse Expression Set 430 | 1429419_PM_at | - | - |
Mouse Expression Set 430 | 1429419_at | - | - |
Murine Genome U74v2 Set | 165495_PM_i_at | - | - |
Murine Genome U74v2 Set | 165495_i_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1429419_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1429419_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 700699 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2940044 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2940044 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002940044 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1222196 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1222196 | - | - |
mouse-6 v1.1 (Illumina) | scl21867.7.1_21-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2940044 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl21867.7.1_21-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2940044 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1222196 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1222196 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl21867.7.1_21-S | - | - |
