Assay Details
Gene Symbol: Dhx34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285931.1 | NP_001272860.1 | 2 - 3 | 1504 | 69 | |
NM_001285932.1 | NP_001272861.1 | 2 - 3 | 1270 | 69 | ||
NM_027883.3 | NP_082159.3 | 2 - 3 | 995 | 69 | ||
GenBank mRNA | AK004733.1 | - | 2 - 3 | 996 | 69 | |
AK076217.1 | - | 1 - 2 | 69 | |||
AK129062.1 | - | 2 - 3 | 978 | 69 | ||
BC058370.1 | - | 2 - 3 | 1332 | 69 | ||
BC080856.1 | - | 2 - 3 | 1270 | 6854931 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
