Assay Details
Gene Symbol: Pan3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028291.4 | NP_082567.3 | 14 - 15 | 2020 | 72 | |
GenBank mRNA | AK012409.1 | - | 3 - 4 | 416 | 72 | |
BC061486.1 | - | 3 - 4 | 613 | 72 | ||
BC082547.1 | - | 13 - 14 | 1549 | 6838444 | 72 | |
BC086321.1 | - | 7 - 8 | 757 | 30108704 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
