Assay Details
| Gene Symbol: Plcd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152813.3 | NP_690026.2 | 11 - 12 | 1942 | 73 | |
| GenBank mRNA | AK011892.1 | - | 7 - 8 | 873 | 73 | |
| AK046669.1 | - | 11 - 12 | 1908 | 73 | ||
| AK144950.1 | - | 3015 | 73 | |||
| AK173311.1 | - | 9 - 10 | 3900 | 73 | ||
| BC031392.1 | - | 11 - 12 | 1886 | 4952407 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plcd3
Genomic Map