Assay Details
Gene Symbol: Slc39a14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135151.1 | NP_001128623.1 | 2 - 3 | 241 | 71 | |
NM_001135152.1 | NP_001128624.1 | 2 - 3 | 207 | 71 | ||
NM_144808.4 | NP_659057.2 | 2 - 3 | 241 | 71 | ||
GenBank mRNA | AB177995.1 | - | 2 - 3 | 244 | 71 | |
AK050076.1 | - | 2 - 3 | 208 | 71 | ||
AK090193.1 | - | 2 - 3 | 206 | 71 | ||
AK122197.1 | - | 563 | 71 | |||
BC021530.1 | - | 2 - 3 | 244 | 5353717 | 71 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AB177995.1,BC021530.1,NM_001135151.1,NM_001135152.1,NM_144808.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
