Assay Details
Gene Symbol: Pear1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032413.1 | NP_001027585.1 | 7 - 8 | 849 | 116 | |
NM_001032414.1 | NP_001027586.1 | 6 - 7 | 779 | 116 | ||
NM_001289600.1 | NP_001276529.1 | 6 - 7 | 751 | 116 | ||
NM_001289601.1 | NP_001276530.1 | 8 - 9 | 992 | 116 | ||
NM_028460.2 | NP_082736.1 | 8 - 9 | 983 | 116 | ||
NM_152799.2 | NP_690012.2 | 8 - 9 | 948 | 116 | ||
GenBank mRNA | AF440279.1 | - | 8 - 9 | 973 | 116 | |
AF444274.1 | - | 6 - 7 | 773 | 116 | ||
AF461685.1 | - | 5 - 6 | 706 | 116 | ||
AK053551.1 | - | 7 - 8 | 840 | 116 | ||
AK141082.1 | - | 746 | 116 | |||
AK161049.1 | - | 8 - 9 | 989 | 116 | ||
BC042490.1 | - | 7 - 8 | 798 | 4953584 | 116 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001289601.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
