Assay Details
Gene Symbol: Tfe3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105196.1 | NP_001098666.1 | 4 - 5 | 1063 | 79 | |
NM_001271491.1 | NP_001258420.1 | 4 - 5 | 783 | 79 | ||
GenBank mRNA | AK089762.1 | - | 3 - 4 | 825 | 79 | |
AK139756.1 | - | 4 - 5 | 1060 | 79 | ||
AK155883.1 | - | 4 - 5 | 785 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
