Assay Details
| Gene Symbol: Vezt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK031848.1 | - | 4 - 5 | 526 | 109 | |
| AK036017.1 | - | 4 - 5 | 474 | 109 | ||
| AK041354.1 | - | 4 - 5 | 503 | 109 | ||
| AK042540.1 | - | 4 - 5 | 468 | 109 | ||
| AY753561.1 | - | 4 - 5 | 430 | 109 | ||
| AY753562.1 | - | 4 - 5 | 430 | 109 | ||
| BC056428.1 | - | 4 - 5 | 461 | 5716245 | 109 | |
| DQ025533.1 | - | 4 - 5 | 430 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vezt
Genomic Map