Assay Details
Gene Symbol: Nr6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159548.1 | NP_001153020.1 | 4 - 5 | 579 | 73 | |
NM_010264.4 | NP_034394.1 | 6 - 7 | 887 | 73 | ||
GenBank mRNA | AF390896.1 | - | 7 - 8 | 1032 | 73 | |
CN537407.1 | - | 4 - 5 | 579 | 73 | ||
CN538225.1 | - | 5 - 6 | 558 | 73 | ||
U09563.1 | - | 6 - 7 | 887 | 73 | ||
U14666.1 | - | 6 - 7 | 846 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
