Assay Details
Gene Symbol: Slc1a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148938.3 | NP_683740.1 | 4 - 5 | 1089 | 67 | |
GenBank mRNA | AF330257.1 | - | 3 - 4 | 589 | 67 | |
AK040798.1 | - | 4 - 5 | 749 | 67 | ||
AK045949.1 | - | 4 - 5 | 768 | 67 | ||
AK049090.1 | - | 4 - 5 | 749 | 67 | ||
AK080879.1 | - | 4 - 5 | 773 | 67 | ||
AK082736.1 | - | 4 - 5 | 749 | 67 | ||
AK084968.1 | - | 4 - 5 | 733 | 67 | ||
AK158777.1 | - | 3 - 4 | 623 | 67 | ||
BC058711.1 | - | 4 - 5 | 1012 | 6842500 | 67 | |
BC066154.1 | - | 4 - 5 | 692 | 30102325 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
