Assay Details
Gene Symbol: Slc38a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175121.3 | NP_780330.2 | 4 - 5 | 697 | 81 | |
GenBank mRNA | AK031615.1 | - | 4 - 5 | 728 | 81 | |
AK129342.1 | - | 4 - 5 | 658 | 81 | ||
BC037675.1 | - | 1 - 2 | 104 | 81 | ||
BC041108.1 | - | 4 - 5 | 567 | 4038546 | 81 | |
BC048178.1 | - | 4 - 5 | 697 | 6825801 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
