Assay Details
| Gene Symbol: Dgkz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166597.1 | NP_001160069.1 | 11 - 12 | 1648 | 101 | |
| NM_138306.2 | NP_612179.2 | 10 - 11 | 1047 | 101 | ||
| GenBank mRNA | AK087503.1 | - | 11 - 12 | 1649 | 101 | |
| AK170936.1 | - | 7 - 8 | 670 | 101 | ||
| BC049228.1 | - | 10 - 11 | 1083 | 3668704 | 101 | |
| BQ443200.1 | - | 1 - 2 | 51 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dgkz
Genomic Map