Assay Details
| Gene Symbol: Dgkz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166597.1 | NP_001160069.1 | 22 - 23 | 2729 | 83 | |
| NM_138306.2 | NP_612179.2 | 21 - 22 | 2128 | 83 | ||
| GenBank mRNA | AK087503.1 | - | 22 - 23 | 2731 | 83 | |
| AK170936.1 | - | 18 - 19 | 1751 | 83 | ||
| AK204403.1 | - | 1 - 2 | 57 | 83 | ||
| BC014860.1 | - | 6 - 7 | 595 | 83 | ||
| BC049228.1 | - | 21 - 22 | 2164 | 3668704 | 83 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AK088841.1 | - | Chr. - - [-] on Build | 83 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Dgkz
Genomic Map