Assay Details
Gene Symbol: Gcg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF276754.1 | - | 3 - 4 | 357 | 100 | |
AK007911.1 | - | 3 - 4 | 350 | 100 | ||
AK148544.1 | - | 3 - 4 | 351 | 100 | ||
BC012975.1 | - | 3 - 4 | 352 | 4218718 | 100 | |
Z46845.1 | - | 2 - 3 | 285 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
