Assay Details
Gene Symbol: Hnrnpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277121.1 | NP_001264050.1 | 2 - 3 | 308 | 89 | |
NM_028871.2 | NP_083147.1 | 2 - 3 | 311 | 89 | ||
GenBank mRNA | AF441128.1 | - | 1 - 2 | 151 | 89 | |
AK141145.1 | - | 2 - 3 | 256 | 89 | ||
AK144682.1 | - | 2 - 3 | 261 | 89 | ||
AK147043.1 | - | 2 - 3 | 277 | 89 | ||
AK152039.1 | - | 2 - 3 | 283 | 89 | ||
BC026850.1 | - | 2 - 3 | 244 | 89 | ||
BC038051.1 | - | 2 - 3 | 288 | 3500728 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
