Assay Details
Gene Symbol: Hnrnpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277121.1 | NP_001264050.1 | 10 - 11 | 1448 | 78 | |
NM_001277122.1 | NP_001264051.1 | 9 - 10 | 1287 | 78 | ||
NM_001277123.1 | NP_001264052.1 | 9 - 10 | 1282 | 78 | ||
NM_028871.2 | NP_083147.1 | 10 - 11 | 1451 | 78 | ||
GenBank mRNA | AF441128.1 | - | 9 - 10 | 1291 | 78 | |
AK046495.1 | - | 3 - 4 | 316 | 78 | ||
AK133840.1 | - | 9 - 10 | 1208 | 78 | ||
AK144682.1 | - | 10 - 11 | 1401 | 78 | ||
AK144992.1 | - | 9 - 10 | 1205 | 78 | ||
AK145849.1 | - | 9 - 10 | 1226 | 78 | ||
AK147043.1 | - | 10 - 11 | 1418 | 78 | ||
BC004679.1 | - | 9 - 10 | 1199 | 78 | ||
BC026850.1 | - | 10 - 11 | 1382 | 78 | ||
BC038051.1 | - | 10 - 11 | 1428 | 3500728 | 78 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
