Assay Details
Gene Symbol: A430057O09 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK079755.1 | - | 8 - 9 | 1229 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 728012 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 559960 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 457912 | AK021323.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 919437 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 20077 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 493610 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 538895 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 850083 | NM_175665.1 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 326385 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 426247 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 864053 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 361830 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 27081 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 696915 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 801601 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 54418 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 518352 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 816972 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 811933 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 287095 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 609724 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 542638 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 410765 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 144565 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 587089 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 884556 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 619583 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 789415 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 826020 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 824490 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 944181 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 531876 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 437519 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 22412 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 465899 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 780421 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 378073 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 315278 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 102606 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 284701 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 963187 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 888042 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 77853 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 34302 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 679644 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 510574 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 89809 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 555195 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 289868 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 568044 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 95356 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 497862 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 41008 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 99690 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 129559 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 33946 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 91088 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 48455 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 17611 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 152719 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 174364 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P100692 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P134640 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P158730 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P134640 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05667884 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05667915 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P158730 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05667893 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05668111 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01168627 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05667873 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05668139 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01168733 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01168691 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05667997 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P100692 | - | - |
Whole Mouse Genome G4122A | A_52_P15532 | AK079755.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P134640 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P158730 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05667997 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P100692 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01168627 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05667873 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04220083 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921549 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921626 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04553875 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921526 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23922164 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23922102 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921574 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921542 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921498 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921576 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921536 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921580 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921513 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921621 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921624 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921592 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921527 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921633 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921567 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921504 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921541 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23922182 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23922169 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23922177 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921495 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04600768 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23922157 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921603 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921530 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921635 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921631 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921600 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04823491 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921525 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921548 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921609 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921619 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04553871 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921602 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04220052 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04981025 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04497030 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23922184 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04823489 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04553885 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921494 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23922111 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921447 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921598 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23922098 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23922107 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921503 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23922103 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23922109 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921448 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921570 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921550 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23922096 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04243444 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23922100 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23922154 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921605 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921601 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04632167 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921506 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04762855 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921545 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921581 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921617 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921509 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04553889 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921608 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23922166 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04220051 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04553883 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921577 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921614 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04275808 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04272039 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921501 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04466288 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921539 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23922172 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04805361 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921623 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921523 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921568 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921534 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921610 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921510 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921583 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921594 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04220088 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23922173 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23922112 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921547 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23922170 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921637 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921611 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04632168 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04981041 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921628 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921524 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23922156 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921533 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23922104 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921521 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23922152 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921613 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23921496 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23922094 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04571808 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 504950 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 1660332 | - | - |
