Assay Details
Gene Symbol: Calcr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042725.1 | NP_001036190.1 | 3 - 4 | 330 | 115 | |
NM_007588.2 | NP_031614.2 | 3 - 4 | 579 | 115 | ||
GenBank mRNA | AK039161.1 | - | 3 - 4 | 524 | 115 | |
AK133983.1 | - | 3 - 4 | 331 | 115 | ||
AK138275.1 | - | 3 - 4 | 581 | 115 | ||
AK162391.1 | - | 3 - 4 | 334 | 115 | ||
BC119232.1 | - | 1 - 2 | 86 | 8733981 | 115 | |
BC119272.1 | - | 1 - 2 | 86 | 8734021 | 115 | |
CJ057218.1 | - | 3 - 4 | 580 | 115 | ||
U18542.1 | - | 3 - 4 | 481 | 115 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK039161.1, AK133983.1, AK138275.1, AK162391.1, BC119232.1, BC119272.1, NM_001042725.1, NM_007588.2, U18542.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
