Assay Details
Gene Symbol: 9330158H04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_015589.1 | 5 - 6 | 667 | 63 | ||
GenBank mRNA | AK034128.1 | - | 5 - 6 | 668 | 63 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | myotrophin | Mtpn |
5033418D15Rik, Gcdp, V1 |
-793.3kb | Sense | Chr. 6 - 35508824 - 35539888 on NCBI Build GRCm38 |
Downstream |
cholinergic receptor, muscarinic 2, cardiac |
Chrm2 |
AChR-M2, Chrm-2, M2 |
51.5kb | Anti-sense | Chr. 6 - 36387950 - 36528638 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2081850 | AK139997.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2081850 | AK139997.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P679586 | AK043142.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125334 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24365198 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366214 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05277783 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366101 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366088 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366218 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24365993 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24365945 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24365184 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24365932 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24365924 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24365930 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24365180 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24365997 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366206 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366097 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24365989 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05489051 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05793635 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366207 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366210 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05001492 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05749285 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24365174 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24365190 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24365940 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366129 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366039 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366036 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366170 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05275361 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366187 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366186 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366046 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366196 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366154 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366188 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05489076 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05793646 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05231979 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24365994 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366151 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366127 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24365196 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366179 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05001499 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366001 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24365919 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366145 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05317898 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366113 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366104 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24365985 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366177 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366045 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366099 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366131 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24365988 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05001466 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366141 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366150 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24365923 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366092 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24365946 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366197 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366079 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24365986 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366103 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05317903 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366159 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366165 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366163 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05870048 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366041 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05793661 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366032 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24365192 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24365987 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366182 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366002 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24365176 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366034 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24365171 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366049 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366193 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05429940 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366044 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366080 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05425012 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366086 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366134 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366000 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05429947 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24365933 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366169 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24365991 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366114 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366089 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05793666 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05277784 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366124 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366212 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366053 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366083 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366118 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24365948 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366156 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05275357 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05429943 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366148 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366095 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05429951 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366175 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366125 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366085 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366178 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366084 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366180 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366166 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366112 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366172 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24365194 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366107 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366116 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366138 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366168 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366142 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05316739 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366087 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24365918 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366183 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366052 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24365925 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366109 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24365921 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05231978 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366082 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366216 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24366100 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.1 ST Array | 10543952 | - | - |
Affymetrix Mouse Clariom S Array | PSR0600016317.mm.1 | TC0600002077.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600016317.mm.1 | NR_015589.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600016317.mm.1 | NR_015589.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 577794 | - | - |
