Assay Details
Gene Symbol: Mmp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008611.4 | NP_032637.3 | 3 - 4 | 608 | 75 | |
GenBank mRNA | AK089234.1 | - | 3 - 4 | 579 | 75 | |
AK089336.1 | - | 3 - 4 | 583 | 75 | ||
AK089381.1 | - | 3 - 4 | 580 | 75 | ||
AK089516.1 | - | 3 - 4 | 569 | 75 | ||
AK137468.1 | - | 3 - 4 | 580 | 75 | ||
AK154937.1 | - | 3 - 4 | 579 | 75 | ||
AK155832.1 | - | 3 - 4 | 579 | 75 | ||
AK155901.1 | - | 3 - 4 | 582 | 75 | ||
AK170177.1 | - | 3 - 4 | 579 | 75 | ||
AK170388.1 | - | 3 - 4 | 581 | 75 | ||
AK170760.1 | - | 3 - 4 | 579 | 75 | ||
AK171168.1 | - | 3 - 4 | 579 | 75 | ||
AK171613.1 | - | 3 - 4 | 581 | 75 | ||
BC042742.1 | - | 3 - 4 | 589 | 5032770 | 75 | |
U96696.1 | - | 3 - 4 | 574 | 75 | ||
Y13342.1 | - | 3 - 4 | 506 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
