Assay Details
Gene Symbol: Mmp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008605.3 | NP_032631.3 | 1 - 2 | 161 | 138 | |
GenBank mRNA | AK089295.1 | - | 130 | 138 | ||
AK089309.1 | - | 1 - 2 | 130 | 138 | ||
AK089335.1 | - | 1 - 2 | 126 | 138 | ||
AK089452.1 | - | 1 - 2 | 130 | 138 | ||
AK089523.1 | - | 1 - 2 | 130 | 138 | ||
AK090051.1 | - | 1 - 2 | 132 | 138 | ||
AK150596.1 | - | 1 - 2 | 130 | 138 | ||
AK151021.1 | - | 1 - 2 | 130 | 138 | ||
AK151273.1 | - | 1 - 2 | 130 | 138 | ||
AK151933.1 | - | 1 - 2 | 130 | 138 | ||
AK155139.1 | - | 1 - 2 | 129 | 138 | ||
AK155743.1 | - | 1 - 2 | 130 | 138 | ||
AK155747.1 | - | 1 - 2 | 130 | 138 | ||
AK155748.1 | - | 1 - 2 | 130 | 138 | ||
AK155865.1 | - | 1 - 2 | 129 | 138 | ||
AK170360.1 | - | 1 - 2 | 130 | 138 | ||
AK170364.1 | - | 1 - 2 | 130 | 138 | ||
AK170798.1 | - | 1 - 2 | 129 | 138 | ||
AK170804.1 | - | 1 - 2 | 129 | 138 | ||
AK170976.1 | - | 1 - 2 | 130 | 138 | ||
AK171006.1 | - | 1 - 2 | 130 | 138 | ||
AK171032.1 | - | 1 - 2 | 129 | 138 | ||
AK171036.1 | - | 1 - 2 | 130 | 138 | ||
AK171176.1 | - | 1 - 2 | 130 | 138 | ||
AK171428.1 | - | 1 - 2 | 130 | 138 | ||
AK171447.1 | - | 1 - 2 | 130 | 138 | ||
AK171448.1 | - | 1 - 2 | 130 | 138 | ||
AK171449.1 | - | 1 - 2 | 130 | 138 | ||
AK171450.1 | - | 1 - 2 | 130 | 138 | ||
AK171466.1 | - | 1 - 2 | 130 | 138 | ||
AK171468.1 | - | 1 - 2 | 130 | 138 | ||
AK171470.1 | - | 1 - 2 | 130 | 138 | ||
AK171472.1 | - | 1 - 2 | 130 | 138 | ||
AK171485.1 | - | 1 - 2 | 130 | 138 | ||
AK171518.1 | - | 1 - 2 | 130 | 138 | ||
AK171542.1 | - | 1 - 2 | 130 | 138 | ||
AK171570.1 | - | 1 - 2 | 130 | 138 | ||
BC019135.1 | - | 1 - 2 | 124 | 5037674 | 138 | |
CT010377.1 | - | 1 - 2 | 77 | 138 | ||
M82831.1 | - | 1 - 2 | 77 | 138 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
