Assay Details
Gene Symbol: Mmp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008605.3 | NP_032631.3 | 8 - 9 | 1263 | 86 | |
GenBank mRNA | AK089309.1 | - | 8 - 9 | 1232 | 86 | |
AK089452.1 | - | 8 - 9 | 1232 | 86 | ||
AK089509.1 | - | 9 - 10 | 1106 | 86 | ||
AK089523.1 | - | 8 - 9 | 1232 | 86 | ||
AK090051.1 | - | 8 - 9 | 1234 | 86 | ||
AK150596.1 | - | 8 - 9 | 1232 | 86 | ||
AK151021.1 | - | 8 - 9 | 1232 | 86 | ||
AK151273.1 | - | 8 - 9 | 1232 | 86 | ||
AK151933.1 | - | 8 - 9 | 1232 | 86 | ||
AK155139.1 | - | 8 - 9 | 1231 | 86 | ||
AK155743.1 | - | 8 - 9 | 1232 | 86 | ||
AK155747.1 | - | 8 - 9 | 1232 | 86 | ||
AK155748.1 | - | 8 - 9 | 1232 | 86 | ||
AK155841.1 | - | 8 - 9 | 1290 | 86 | ||
AK155865.1 | - | 8 - 9 | 1231 | 86 | ||
AK170360.1 | - | 8 - 9 | 1232 | 86 | ||
AK170364.1 | - | 8 - 9 | 1232 | 86 | ||
AK170500.1 | - | 9 - 10 | 1106 | 86 | ||
AK170798.1 | - | 8 - 9 | 1231 | 86 | ||
AK170804.1 | - | 8 - 9 | 1231 | 86 | ||
AK170976.1 | - | 8 - 9 | 1232 | 86 | ||
AK171006.1 | - | 8 - 9 | 1232 | 86 | ||
AK171032.1 | - | 8 - 9 | 1231 | 86 | ||
AK171036.1 | - | 8 - 9 | 1232 | 86 | ||
AK171176.1 | - | 8 - 9 | 1232 | 86 | ||
AK171428.1 | - | 8 - 9 | 1232 | 86 | ||
AK171435.1 | - | 7 - 8 | 2279 | 86 | ||
AK171447.1 | - | 8 - 9 | 1232 | 86 | ||
AK171448.1 | - | 8 - 9 | 1232 | 86 | ||
AK171449.1 | - | 8 - 9 | 1232 | 86 | ||
AK171450.1 | - | 8 - 9 | 1232 | 86 | ||
AK171466.1 | - | 8 - 9 | 1232 | 86 | ||
AK171468.1 | - | 8 - 9 | 1232 | 86 | ||
AK171470.1 | - | 8 - 9 | 1232 | 86 | ||
AK171472.1 | - | 8 - 9 | 1232 | 86 | ||
AK171485.1 | - | 8 - 9 | 1232 | 86 | ||
AK171518.1 | - | 8 - 9 | 1232 | 86 | ||
AK171542.1 | - | 8 - 9 | 1232 | 86 | ||
AK171570.1 | - | 8 - 9 | 1232 | 86 | ||
M82831.1 | - | 8 - 9 | 1179 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
