Assay Details
Gene Symbol: Ddx50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053183.2 | NP_444413.1 | 1 - 2 | 170 | 77 | |
GenBank mRNA | AF334104.1 | - | 1 - 2 | 171 | 77 | |
BC049906.1 | - | 1 - 2 | 172 | 77 | ||
BC055440.1 | - | 1 - 2 | 164 | 77 | ||
BC075714.1 | - | 1 - 2 | 122 | 77 | ||
BC090996.1 | - | 1 - 2 | 190 | 6757408 | 77 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
