Assay Details
Gene Symbol: Phactr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033257.4 | NP_001028429.1 | 3 - 4 | 343 | 110 | |
NM_001195065.1 | NP_001181994.1 | 3 - 4 | 587 | 110 | ||
NM_001195066.1 | NP_001181995.1 | 3 - 4 | 522 | 110 | ||
NM_001195096.1 | NP_001182025.1 | 3 - 4 | 343 | 110 | ||
GenBank mRNA | AK139466.1 | - | 3 - 4 | 523 | 110 | |
AK142930.1 | - | 3 - 4 | 344 | 110 | ||
BC012871.1 | - | 3 - 4 | 611 | 110 | ||
BC130264.1 | - | 3 - 4 | 339 | 40142964 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
