Assay Details
| Gene Symbol: Pole4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025882.3 | NP_080158.1 | 2 - 3 | 329 | 75 | |
| GenBank mRNA | AK010135.1 | - | 2 - 3 | 324 | 75 | |
| AK010300.1 | - | 2 - 3 | 330 | 75 | ||
| AK011595.1 | - | 2 - 3 | 341 | 75 | ||
| AK145365.1 | - | 2 - 3 | 335 | 75 | ||
| AK160582.1 | - | 2 - 3 | 341 | 75 | ||
| AK167631.1 | - | 2 - 3 | 319 | 75 | ||
| BC023189.1 | - | 2 - 3 | 307 | 4952949 | 75 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pole4
Genomic Map