Assay Details
| Gene Symbol: Tex261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009357.2 | NP_033383.1 | 2 - 3 | 239 | 69 | |
| GenBank mRNA | AK149789.1 | - | 2 - 3 | 204 | 69 | |
| AK157834.1 | - | 2 - 3 | 218 | 69 | ||
| BC003802.1 | - | 2 - 3 | 183 | 3486968 | 69 | |
| BY250528.1 | - | 2 - 3 | 240 | 69 | ||
| X81058.1 | - | 2 - 3 | 185 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tex261
Genomic Map